Heptelidic Acid

Synonyms: AvocettinKoningic acidFO-4443BRN 5091359

2215-250 2215-1000
10189-214EA 180.96 USD
10189-214 10189-216
Heptelidic Acid
Heptelidic Acid
Formula: C₁₅H₂₀O₅
MW: 280.32 g/mol
CAS Number: 74310-84-2

Order Now

Specification Test Results

CAS 74310-84-2
Purity ³95% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR