10Z-Hymenialdisine

Synonyms: Hymenialdisine

2212-1000 2212-250
10189-202EA 204.31 USD
10189-202 10189-204
10Z-Hymenialdisine
10Z-Hymenialdisine
Formula: C₁₁H₁₀BrN₅O₂
MW: 324.1 g/mol
CAS Number: 82005-12-7

Order Now

Specification Test Results

CAS 82005-12-7
Purity ≥97% by HPLC
Form Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR