Pyroxamide

Supplier: BioVision

AB144633-5MG AB144633-25MG
10189-132EA 804.39 USD
10189-132 10189-130
Pyroxamide
Pyroxamide
CAS Number: 382180-17-8

Order Now

Specification Test Results

CAS 382180-17-8
Purity ≥95%
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR