Nullscript

Synonyms: 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide

2186-5 2186-1
10189-118EA 116.75 USD
10189-118 10189-120
Nullscript
Nullscript
Formula: C₁₆H₁₄N₂O₄
MW: 298.30 g/mol
MDL Number: MFCD00405081
CAS Number: 300816-11-9

Order Now

Specification Test Results

CAS 300816-11-9
Purity ≥97% by TLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR