Latrunculin A

Synonyms: (4R)-4-[(1R,4S,5E,7Z,11E,15R,17R)-17-hydroxy-4,11-dimethyl-13-oxo-14,18-dioxabicyclo[13.3.1]nonadeca-5,7,11-trien-17-yl]-1,3-thiazolidin-2-one

2181-100
10189-102EA 219.09 USD
10189-102
Latrunculin A
Latrunculin A
Formula: C₂₂H₃₁NO₅S
MW: 421.552 g/mol
CAS Number: 76343-93-6

Order Now

Specification Test Results

CAS 76343-93-6
Purity ≥96% by HPLC
Form White to light yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR