Ebselen

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

2169-5 2169-25
10189-074EA 262.69 USD
10189-074 10189-072
Ebselen
Ebselen
Formula: C₁₃H₉NOSe
MW: 274.20 g/mol
MDL Number: MFCD00210937
CAS Number: 60940-34-3

Order Now

Specification Test Results

CAS 60940-34-3
Purity ≥99%
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR