Milrinone

Synonyms: 6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone2-Methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile

2163-25 2163-5
10191-320EA 140.1 USD
10191-320 10192-022
Milrinone
Milrinone
Formula: C₁₂H₉N₃O
MW: 211.22 g/mol
MDL Number: MFCD00133539
CAS Number: 78415-72-2

Order Now

Specification Test Results

CAS 78415-72-2
Purity ≥98%
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR