Direct blue 1

Synonyms: 6,6'-((3,3'-Dimethoxy-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(4-amino-5-hydroxynaphthalene-1,3-disulfonic acid), tetrasodium salt6,6'-((3,3'-Dimethoxy-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(4-amino-5-hydroxynaphthalene-1,3-disulphonic acid), tetrasodium saltChicago Sky Blue 6BPontamine Sky Blue 6B

2158-1G
10189-042EA 81.72 USD
10189-042
Direct blue 1
Direct blue 1
Formula: C₃₄H₂₈N₆O₁₆S₄
MW: 992.79 g/mol
MDL Number: MFCD00004020
CAS Number: 2610-05-1

Order Now

Specification Test Results

CAS 2610-05-1
Purity Dye content: ~ 80%
Form Brown to dark brown solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR