Hyperforin DCHA

2153-1000 2153-500
10189-024EA 332.74 USD
10189-024 10189-022
Hyperforin DCHA
Hyperforin DCHA
Formula: C₃₅H₅₁O₄·C₁₂H₂₄N
MW: 535.8·182.3 g/mol
CAS Number: 238074-03-8

Order Now

Specification Test Results

CAS 238074-03-8
Purity ≥97% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR