Amidepsine A (from Humcola sp.)
2147-1000
2147-500
10189-002EA
484.51
USD
10189-002
10189-000
Amidepsine A (from Humcola sp.)
Amidepsine A
Formula:
C₂₉H₂₉NO₁₁ |
CAS Number:
169181-28-6 |
Specification Test Results
CAS | 169181-28-6 |
Purity | ≥95% by HPLC |
Form | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...