BAY 43-9006 free base

Synonyms: N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)phenoxy)-N-methylpicolinamide4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide

2142-100 2142-25
10188-988EA 128.42 USD
10188-988 10188-990
BAY 43-9006 free base
Sorafenib (BAY 43-9006)
Formula: C₂₁H₁₆ClF₃N₄O₃
MW: 464.82 g/mol
MDL Number: MFCD06411450
CAS Number: 284461-73-0

Order Now

Specification Test Results

CAS 284461-73-0
Purity ≥99% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR