Nonidet® P 40 Substitute (NP-40) 10% in aqueous solution, MegaPure™ detergent

Supplier: BioVision

2127-500 2127-SET 2127-50 2127-100
10188-938EA 625.93 USD
10188-938 76289-910 10191-310 10188-936
Nonidet® P 40 Substitute (NP-40) 10% in aqueous solution, MegaPure™ detergent
Nonidet® P 40 Substitute (NP-40)

A non-ionic surfactant, used to solubilize cerebral GABA receptors and for solubilization of proteins and lipids of the scallop gill ciliary membrane and of membranes from breast cancer tumors.


  • A non-ionic detergent

Order Now

Specification Test Results

CAS 9016-45-9
Purity Aldehydes: 200 µM; Peroxides: 200 µM
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR