Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate solution 100 mM
Supplier: BioVision
AB181719-100UL
10191-288EA
99.34
USD
10191-288
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate solution 100 mM
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate
Formula:
C₁₀H₁₄N₅Na₂O₁₃P₃·H₂O |
MDL Number:
MFCD00150755 CAS Number: 34369-07-8 |
Specification Test Results
CAS | 34369-07-8 |
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...