Decylubiquinone
2099-10
2099-1
10188-930EA
180.68
USD
10188-930
10188-928
Decylubiquinone
Decylubiquinone
CAS Number:
55486-00-5 |
Specification Test Results
CAS | 55486-00-5 |
Purity | ≥98% |
Form | Orange liquid or semisolid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...