2-Chloro-5-nitro-N-(4-pyridyl)benzamide

Synonyms: 2-Chloro-5-nitro-N-(pyridin-4-yl)benzamide

2095-5
10188-914EA 140.1 USD
10188-914
2-Chloro-5-nitro-N-(4-pyridyl)benzamide
2-Chloro-5-nitro-N-(4-pyridyl)benzamide
Formula: C₁₂H₈ClN₃O₃
MW: 277.7 g/mol
MDL Number: MFCD00121849
CAS Number: 313516-66-4

Order Now

Specification Test Results

CAS 313516-66-4
Purity ≥98% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR