LDN193189, EZSolution™

Synonyms: 4-(6-(4-(Piperazin-1-yl)phenyl)pyrazolo[1,5-a]pyrimidin-3-yl)quinoline

2092-5
10188-906EA 266.9 USD
10188-906
LDN193189, EZSolution™
LDN193189
Formula: C₂₅H₂₂N₆
MW: 406,48 g/mol
MDL Number: MFCD17392570
CAS Number: 1062368-24-4

Order Now

Specification Test Results

CAS 1062368-24-4 (free base)
Purity ≥97% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR