Garcinol

Supplier: BioVision

AB141503-25MG AB141503-5MG
10188-894EA 727.81 USD
10188-894 10188-892
Garcinol
Garcinol
CAS Number: 78824-30-3

Order Now

Specification Test Results

CAS 78824-30-3
Purity ≥95% by TLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR