B2

Synonyms: (4-Chlorophenyl)(4-(8-nitroquinolin-5-yl)piperazin-1-yl)methanone

2085-5
10188-884EA 204.31 USD
10188-884
B2
B2
Formula: C₂₀H₁₇ClN₄O₃
MW: 396.8 g/mol
MDL Number: MFCD02039810
CAS Number: 115687-05-3

Order Now

Specification Test Results

CAS 115687-05-3
Purity ≥95%
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR