Digitonin, water soluble, EZSolution™

Supplier: BioVision

AB300022-1ML
10191-280EA 214.21 USD
10191-280
Digitonin, water soluble, EZSolution™
Digitonin
Formula: C₅₆H₉₂O₂₉
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

CAS 11024-24-1
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR