Pterostilbene ≥98%

Synonyms: (E)-4-(3,5-Dimethoxystyryl)phenol

2073-25 2073-5
10188-862EA 326.9 USD
10188-862 10188-860
Pterostilbene ≥98%
Pterostilbene
Formula: C₁₆H₁₆O₃
MW: 256.2964 g/mol
MDL Number: MFCD00238710
CAS Number: 537-42-8

Order Now

Specification Test Results

CAS 537-42-8
Purity ≥98% by LC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR