Piceatannol

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

2072-5 2072-25
10188-856EA 81.72 USD
10188-856 10188-858
Piceatannol
Piceatannol
Formula: C₁₄H₁₂O₄
MW: 244.2427 g/mol
MDL Number: MFCD00221715
CAS Number: 10083-24-6

Order Now

Specification Test Results

CAS 10083-24-6
Purity ≥98% by LC
Form Off-white to light pink solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR