PK 11195

2054-10 2054-50
10191-264EA 81.72 USD
10191-264 10188-806
PK 11195
PK 11195
Formula: C₂₁H₂₁ClN₂O
CAS Number: 85532-75-8

Order Now

Specification Test Results

CAS 85532-75-8
Purity ≥98% by NMR
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR