Pellitorine ≥97% (by HPLC)

Supplier: BioVision

Synonyms: (2E,4E)-N-Isobutyldeca-2,4-dienamide

AB141495-5MG AB141495-1MG
10191-260EA 268.11 USD
10191-260
Pellitorine ≥97% (by HPLC)
Pellitorine

Pellitorine is a natural product isolated from the roots of piper nigrum.


  • Cell-permeable
  • Modulator of the sensory neuron function to induce tingling parethesia
  • Excellent stable model compound for sensory studies


α-Glucosidase inhibitor used in diabetes mellitus, cancer, infection and inflammatory research. Also acts as a ACAT (Acyl-CoA cholesteryl acyl transferase) inhibitor. Potential anti-cancer lead compound.


Caution: For Research Use Only! Not For Use in Humans.

Formula: C₁₄H₂₅NO
MW: 223.4 g/mol
MDL Number: MFCD01735995
CAS Number: 18836-52-7

Order Now

Specification Test Results

CAS 18836-52-7
Purity ≥97% by HPLC
Form Yellow to beige solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR