Skyrin (from Talaromyces sp.)

Supplier: BioVision

Synonyms: EndothianinRhodophyscin

AB142207-1MG
10188-784EA 589.9 USD
10188-784
Skyrin (from Talaromyces sp.)
Skyrin
Formula: C₃₀H₁₈O₁₀
MW: 538.46
CAS Number: 602-06-2

Order Now

Specification Test Results

CAS 602-06-2
Purity ≥97% by HPLC
Form Orange to dark red solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR