RO 20-1724

Supplier: BioVision

AB141492-250MG AB141492-50MG
10188-780EA 168.28 USD
10188-780 10188-782
RO 20-1724
RO 20-1724
Formula: C₁₅H₂₂N₂O₃
MW: 278.3 g/mol
CAS Number: 29925-17-5

Order Now

Specification Test Results

CAS 29925-17-5
Purity ≥99% by TLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR