GX15-070
Specification Test Results
CAS | 803712-67-6 |
Purity | ≥98% |
Form | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
CAS | 803712-67-6 |
Purity | ≥98% |
Form | Off-white solid |
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
© 2024 VWR International, LLC. All rights reserved.
© 2024 FORTUNE Media IP Limited All rights reserved. Used under license.