Spautin-1

Supplier: BioVision

Synonyms: 6-Fluoro-N-(4-fluorobenzyl)quinazolin-4-amine6-Fluoro-N-[(4-fluorophenyl)methyl]-4-quinazolinamine

AB142205-5MG AB142205-25MG
10188-766EA 1394.26 USD
10188-766 10188-764
Spautin-1
Spautin-1
Formula: C₁₅H₁₁F₂N₃
MW: 271.27 g/mol
MDL Number: MFCD16962292
CAS Number: 1262888-28-7

Order Now

Specification Test Results

CAS 1262888-28-7
Purity ≥97% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR