n-Dodecyl-β-D-maltoside

Synonyms: Dodecyl-β-D-maltopyranoside(2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triolLauryl-β-D-maltosideDDM

2036-25 2036-50 2036-5 2036-1
10188-758EA 521.17 USD
10188-758 10188-762 10188-760 10191-258
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now

Specification Test Results

CAS 69227-93-6
Purity ≥99% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR