b-AP15 [DUB inhibitor] solution

Supplier: BioVision

Synonyms: VLX1500(3E,5E)-1-Acryloyl-3,5-bis(4-nitrobenzylidene)piperidin-4-one

AB142195-5MG AB142195-25MG
10191-738EA 191.54 USD
10191-738 10191-740
b-AP15 [DUB inhibitor] solution
b-AP15 [DUB inhibitor] solution
Formula: C₂₂H₁₇N₃O₆
MW: 419.4 g/mol
MDL Number: MFCD26142662
CAS Number: 1009817-63-3

Order Now

Specification Test Results

CAS 1009817-63-3
Purity ≥98% by HPLC
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR