(+/-)-Thalidomide

Synonyms: N-(2,6-Dioxo-3-piperidinyl)phthalimide2-(2,6-Dioxopiperidin-3-yl)isoindoline-1,3-dione

2020-50 2020-10
10188-734EA 81.6 USD
10188-734 10188-736
(+/-)-Thalidomide
(+/-)-Thalidomide
Formula: C₁₃H₁₀N₂O₄
MW: 258.23 g/mol
MDL Number: MFCD00153873
CAS Number: 50-35-1

Order Now

Specification Test Results

CAS 50-35-1
Purity ≥98% by TLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR