ZM 447439

Synonyms: N-(4-((6-Methoxy-7-(3-morpholinopropoxy)quinazolin-4-yl)amino)phenyl)benzamide

2018-5 2018-1
10188-726EA 93.39 USD
10188-726 10188-728
ZM 447439
ZM 447439
Formula: C₂₉H₃₁N₅O₄
MW: 513.6 g/mol
MDL Number: MFCD08703125
CAS Number: 331771-20-1

Order Now

Specification Test Results

CAS 331771-20-1
Purity ≥98% by HPLC
Form Orange solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR