WP-1130

Synonyms: (S,E)-3-(6-Bromopyridin-2-yl)-2-cyano-N-(1-phenylbutyl)acrylamide

2014-5 2014-25
10188-716EA 93.26 USD
10188-716 10188-718
WP-1130
WP-1130
Formula: C₁₉H₁₈BrN₃O
MW: 384,27 g/mol
MDL Number: MFCD18839228
CAS Number: 856243-80-6

Order Now

Specification Test Results

CAS 856243-80-6
Purity ≥98% by TLC
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR