K252a
2013-1000
2013-500
10188-712EA
105.07
USD
10188-712
10188-714
K252a
K252a
CAS Number:
99533-80-9 |
Specification Test Results
CAS | 99533-80-9 |
Purity | ≥98% by HPLC |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...