RO4929097

Synonyms: (S)-2,2-Dimethyl-N1-(6-oxo-6,7-dihydro-5H-dibenzo[b,d]azepin-7-yl)-N3-(2,2,3,3,3-pentafluoropropyl)malonamide

2011-5 2011-1
10192-020EA 503.18 USD
10192-020 10192-018
RO4929097
RO4929097
Formula: C₂₂H₂₀F₅N₃O₃
MW: 469,4 g/mol
MDL Number: MFCD18384976
CAS Number: 847925-91-1

Order Now

Specification Test Results

CAS 847925-91-1
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR