Y-27632, EZSolution™, sterile filtered

Supplier: BioVision

Synonyms: (R)-(+)-trans-N-(4-Pyridyl)-4-(1-aminoethyl)-cyclohexanecarboxamide, 2HCl(R)-(+)-trans-4-(1-Aminoethyl)-N-(4-Pyridyl)cyclohexanecarboxamide dihydrochlorideROCK InhibitorY-27632 dihydrochlorideRho Kinase Inhibitor VI

1994-1
10191-234EA 320.76 USD InStock
10191-234
Y-27632, EZSolution™, sterile filtered
Y-27632
Formula: C₁₄H₂₁N₃O·2HCl
MW: 320,26 g/mol
MDL Number: MFCD03490488
CAS Number: 146986-50-7

Order Now

Specification Test Results

CAS 146986-50-7
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR