CHIR99021, EZSolution™, sterile filtered

Supplier: BioVision

Synonyms: 6-((2-((4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl)amino)ethyl)amino)nicotinonitrileGSK-3 Inhibitor XVIAGN-PC-0MWAEXCHEMBL412142

AB291107-1ML
10188-676EA 554.41 USD
10188-676
CHIR99021, EZSolution™, sterile filtered
CHIR99021
Formula: C₂₂H₁₈Cl₂N₈
MW: 465.3379 g/mol
MDL Number: MFCD11846251
CAS Number: 252917-06-9

Order Now

Specification Test Results

CAS 252917-06-9
Purity ≥95% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR