(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide, EZSolution™, sterile filtered

Supplier: BioVision

Synonyms: (R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamidePD-0325901

AB289849-1ML
10188-674EA 477.83 USD
10188-674
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide, EZSolution™, sterile filtered
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide
Formula: C₁₆H₁₄F₃IN₂O₄
MW: 482.2 g/mol
MDL Number: MFCD08435926
CAS Number: 391210-10-9

Order Now

Specification Test Results

CAS 391210-10-9
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR