Tubacin

Synonyms: rel-N1-(4-((2R,4R,6S)-4-(((4,5-Diphenyloxazol-2-yl)thio)methyl)-6-(4-(hydroxymethyl)phenyl)-1,3-dioxan-2-yl)phenyl)-N8-hydroxyoctanediamide

1984-250 1984-1000
10188-648EA 105.07 USD
10188-648 10188-650
Tubacin
Tubacin
Formula: C₄₁H₄₃N₃O₇S
MW: 721,86 g/mol
MDL Number: MFCD28144524
CAS Number: 537049-40-4

Order Now

Specification Test Results

CAS 537049-40-4
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR