CID 2745687

Supplier: BioVision

AB141477-25MG AB141477-5MG
10188-610EA 1287.01 USD
10188-610 10188-608
CID 2745687
CID 2745687
CAS Number: 264233-05-8

Order Now

Specification Test Results

CAS 264233-05-8
Purity ≥95% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR