PX-866
1965-1
1965-5
10191-206EA
320.58
USD
10191-206
10191-204
PX-866
PX-866
CAS Number:
502632-66-8 |
Specification Test Results
CAS | 502632-66-8 |
Purity | ≥98% by TLC |
Form | Orange solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...