NVP DPP 728

Supplier: BioVision

AB142171-5MG AB142171-1MG
10191-200EA 222.16 USD
10191-200 10191-202
NVP DPP 728
NVP DPP 728
CAS Number: 247016-69-9

Order Now

Specification Test Results

CAS 247016-69-9
Purity ≥98% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR