K 579
1963-1
1963-5
10191-198EA
461.16
USD
10191-198
10191-196
K 579
K 579
CAS Number:
440100-64-1 |
Specification Test Results
CAS | 440100-64-1 |
Purity | ≥98% by HPLC |
Form | Light brown solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...