Vinorelbine tartrate

Supplier: BioVision

AB142167-5MG AB142167-25MG
10191-728EA 162.35 USD
10191-728 10191-730
Vinorelbine tartrate
Vinorelbine tartrate
CAS Number: 125317-39-7

Order Now

Specification Test Results

CAS 125317-39-7
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR