GSK-3beta inhibitor VIII

Synonyms: 1-(4-Methoxybenzyl)-3-(5-nitrothiazol-2-yl)ureaN-(4-Methoxybenzyl)-N′-(5-nitro-1,3-thiazol-2-yl)ureaAR-A014418

1953-2 1953-5
10188-586EA 81.6 USD
10188-586 10188-588
GSK-3beta inhibitor VIII
GSK-3beta inhibitor VIII
Formula: C₁₂H₁₂N₄O₄S
MW: 308.31 g/mol
MDL Number: MFCD08277040
CAS Number: 487021-52-3

Order Now

Specification Test Results

CAS 487021-52-3
Purity ≥98% by TLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR