(R)-Roscovitine
Synonyms:
Roscovitine
, 2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-benzylamino-9-isopropylpurine
1950-1
1950-5
10191-188EA
227.66
USD
10191-188
10191-186
(R)-Roscovitine
(R)-Roscovitine
Formula:
C₁₉H₂₆N₆O MW: 354.45 g/mol |
MDL Number:
MFCD02266401 CAS Number: 186692-46-6 |
Specification Test Results
CAS | 186692-46-6 |
Purity | ≥98% by HPLC |
Form | White to off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...