C646

Synonyms: 4-(4-((5-(4,5-Dimethyl-2-nitrophenyl)furan-2-yl)methylene)-3-methyl-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid

1948-1 1948-5
10191-178EA 105.07 USD
10191-178 10191-180
C646
C646
Formula: C₂₄H₁₉N₃O₆
MW: 445.4 g/mol
MDL Number: MFCD01784780
CAS Number: 328968-36-1

Order Now

Specification Test Results

CAS 328968-36-1
Purity ≥98% by NMR
Form Red solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR