Celastrol

Synonyms: Tripterin

1940-5 1940-25
10188-554EA 286.04 USD
10188-554 10188-552
Celastrol
Celastrol
Formula: C₂₉H₃₈O₄
MW: 450.62 g/mol
MDL Number: MFCD03424073
CAS Number: 34157-83-0

Order Now

Specification Test Results

CAS 34157-83-0
Purity ≥98% by TLC
Form Red solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR