SU 11274

Supplier: BioVision

AB142159-25MG AB142159-5MG
10188-546EA 1393.89 USD
10188-546 10188-544
SU 11274
SU 11274
CAS Number: 658084-23-2

Order Now

Specification Test Results

CAS 658084-23-2
Purity ≥98% by HPLC
Form Orange solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR