UNC0638

Supplier: BioVision

Synonyms: 2-Cyclohexyl-N-(1-isopropylpiperidin-4-yl)-6-methoxy-7-(3-(pyrrolidin-1-yl)propoxy)quinazolin-4-amine

AB142157-1MG AB142157-5MG
10188-530EA 1170.35 USD
10188-530 10188-528
UNC0638
UNC0638
Formula: C₃₀H₄₇N₅O₂
MW: 509,73 g/mol
MDL Number: MFCD22208587
CAS Number: 1255580-76-7

Order Now

Specification Test Results

CAS 1255580-76-7
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR