PYR-41

Supplier: BioVision

Synonyms: Ethyl 4-(4-((5-nitrofuran-2-yl)methylene)-3,5-dioxopyrazolidin-1-yl)benzoate

AB141469-5MG
10191-706EA 873.36 USD
10191-706 10191-704
PYR-41
PYR-41
Formula: C₁₇H₁₃N₃O₇·3H₂O
MW: 425.40 g/mol
MDL Number: MFCD01469983
CAS Number: 418805-02-4

Order Now

Specification Test Results

CAS 418805-02-4
Purity ≥98% by TLC
Form Brown solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR